The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005024.2 | NP_005015.1 | 2 - 3 | 231 | 108 |
XM_011526027.1 | XP_011524329.1 | 3 - 4 | 326 | 108 | |
XM_017025793.1 | XP_016881282.1 | 2 - 3 | 231 | 108 | |
XM_017025794.1 | XP_016881283.1 | 2 - 3 | 231 | 108 | |
GenBank mRNA | AK314936.1 | - | 2 - 3 | 231 | 108 |
BC096217.3 | - | 1 - 2 | 171 | 108 | |
BC096218.3 | - | 1 - 2 | 171 | 108 | |
BC096219.3 | - | 1 - 2 | 171 | 108 | |
BC096220.3 | - | 1 - 2 | 171 | 108 | |
U35459.1 | - | 1 - 2 | 171 | 108 |