The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018045.7 | NP_060515.3 | 3 - 4 | 235 | 89 |
NR_125352.1 | 3 - 4 | 235 | 89 | ||
XM_005270986.4 | XP_005271043.1 | 3 - 4 | 215 | 89 | |
GenBank mRNA | AK001138.1 | - | 3 - 4 | 205 | 89 |
AK021440.1 | - | 3 - 4 | 214 | 89 | |
AK225026.1 | - | 3 - 4 | 216 | 89 | |
AK225717.1 | - | 3 - 4 | 205 | 89 | |
AK297352.1 | - | 3 - 4 | 214 | 89 | |
BC010573.1 | - | 3 - 4 | 194 | 89 | |
BC026322.1 | - | 2 - 3 | 202 | 89 | |
BC028381.2 | - | 3 - 4 | 207 | 89 | |
BP363955.1 | - | 3 - 4 | 235 | 89 | |
CR457252.1 | - | 2 - 3 | 188 | 89 |