The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021020.3 | NP_066300.1 | 1 - 2 | 460 | 56 |
XM_005273394.3 | XP_005273451.1 | 2 - 3 | 725 | 56 | |
XM_011544383.2 | XP_011542685.1 | 2 - 3 | 2663 | 56 | |
XM_011544384.2 | XP_011542686.1 | 2 - 3 | 613 | 56 | |
XM_011544385.2 | XP_011542687.1 | 2 - 3 | 587 | 56 | |
XM_011544386.2 | XP_011542688.1 | 2 - 3 | 812 | 56 | |
GenBank mRNA | AF123654.1 | - | 1 - 2 | 349 | 56 |
AF123655.1 | - | 1 - 2 | 349 | 56 | |
AF123656.1 | - | 1 - 2 | 349 | 56 | |
AF123657.1 | - | 1 - 2 | 349 | 56 | |
AF123658.1 | - | 1 - 2 | 349 | 56 | |
AF123659.1 | - | 1 - 2 | 460 | 56 | |
BC075006.2 | - | 1 - 2 | 398 | 56 | |
BC075007.2 | - | 1 - 2 | 398 | 56 |