Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP1R3D
Entrez Gene ID
5509
Gene Name
protein phosphatase 1 regulatory subunit 3D
Gene Aliases
PPP1R6
Chromosome Location
Chr.20: 59936832 - 59940297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006242.3 NP_006233.1 1 - 1 329 86
GenBank mRNA BC047502.1 - 1 - 1 329 86
Y18206.1 - 1 - 1 239 86

View Full Product Details