The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK091344.1 | - | 1 - 1 | 1183 | 55 |
AK299614.1 | - | 3 - 3 | 817 | 55 | |
AK315589.1 | - | 1 - 1 | 1178 | 55 | |
AL136782.1 | - | 1 - 1 | 1095 | 55 | |
BC022033.2 | - | 1 - 1 | 1178 | 55 | |
CR533558.1 | - | 1 - 1 | 869 | 55 | |
FJ150424.1 | - | 1 - 1 | 1178 | 55 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152903.4 | NP_690867.3 | 1 - 1 | 1158 | 55 |
GenBank mRNA | AK056633.1 | - | 1 - 1 | 1103 | 55 |
AK096608.1 | - | 2 - 2 | 1098 | 55 | |
AK299627.1 | - | 2 - 2 | 906 | 55 | |
AL833839.1 | - | 1 - 1 | 810 | 55 | |
BC000560.2 | - | 1 - 1 | 1080 | 55 | |
BC051349.1 | - | 1 - 1 | 919 | 55 |