The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199237.1 | NP_001186166.1 | 3 - 4 | 1315 | 79 |
NM_001199238.1 | NP_001186167.1 | 3 - 4 | 1184 | 79 | |
NM_001199239.1 | NP_001186168.1 | 3 - 4 | 1181 | 79 | |
NM_001199240.1 | NP_001186169.1 | 3 - 4 | 967 | 79 | |
NM_033177.3 | NP_149417.1 | 2 - 3 | 1291 | 79 | |
XM_005249403.3 | XP_005249460.1 | 3 - 4 | 1318 | 79 | |
XM_006715204.1 | XP_006715267.1 | 3 - 4 | 996 | 79 | |
XM_011514909.1 | XP_011513211.1 | 3 - 4 | 1036 | 79 | |
XM_011514910.1 | XP_011513212.1 | 3 - 4 | 1033 | 79 | |
GenBank mRNA | AJ420471.1 | - | 2 - 3 | 883 | 79 |
AK123707.1 | - | 3 - 4 | 1315 | 79 | |
AK314056.1 | - | 3 - 4 | 1184 | 79 | |
BC008783.1 | - | 2 - 3 | 1281 | 79 | |
BC018875.2 | - | 2 - 3 | 1275 | 79 |