The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004982.3 | NP_004973.1 | 1 - 2 | 281 | 100 |
XM_005253358.4 | XP_005253415.1 | 2 - 3 | 250 | 100 | |
XM_017019283.1 | XP_016874772.1 | 2 - 3 | 185 | 100 | |
XM_017019284.1 | XP_016874773.1 | 2 - 3 | 256 | 100 | |
GenBank mRNA | AK291821.1 | - | 2 - 3 | 89 | 100 |
AL546327.3 | - | 1 - 2 | 281 | 100 | |
BC000544.2 | - | 1 - 2 | 141 | 100 | |
D50312.1 | - | 1 - 2 | 205 | 100 |