The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005683.3 | NP_005674.2 | 2 - 2 | 311 | 80 |
XM_005246952.3 | XP_005247009.1 | 2 - 2 | 571 | 80 | |
XM_011512175.2 | XP_011510477.1 | 2 - 2 | 747 | 80 | |
XM_011512176.2 | XP_011510478.1 | 2 - 2 | 330 | 80 | |
GenBank mRNA | AK292223.1 | - | 2 - 2 | 456 | 80 |
BC032694.1 | - | 2 - 2 | 311 | 80 | |
CR541776.1 | - | 1 - 1 | 118 | 80 | |
FJ348259.1 | - | 1 - 1 | 118 | 80 |