The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168364.1 | NP_001161836.1 | 5 - 6 | 664 | 88 |
NM_001321325.1 | NP_001308254.1 | 5 - 6 | 664 | 88 | |
NM_173853.3 | NP_776252.2 | 5 - 6 | 664 | 88 | |
GenBank mRNA | AY157576.1 | - | 5 - 6 | 634 | 88 |
AY358993.1 | - | 4 - 5 | 722 | 88 | |
BC101687.1 | - | 5 - 6 | 663 | 88 | |
BC101689.1 | - | 5 - 6 | 664 | 88 | |
BC143574.1 | - | 5 - 6 | 663 | 88 | |
BM794111.1 | - | 1 - 2 | 83 | 88 | |
CB160871.1 | - | 2 - 3 | 88 |