The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130020.1 | NP_001123492.1 | 21 - 21 | 3091 | 99 |
NM_001130021.1 | NP_001123493.1 | 22 - 22 | 3088 | 99 | |
NM_005177.3 | NP_005168.2 | 21 - 21 | 3070 | 99 | |
XM_005257459.1 | XP_005257516.1 | 22 - 22 | 3091 | 99 | |
XM_005257461.1 | XP_005257518.1 | 21 - 21 | 2919 | 99 | |
XM_005257463.2 | XP_005257520.1 | 20 - 20 | 2901 | 99 | |
XM_011524908.1 | XP_011523210.1 | 20 - 20 | 2965 | 99 | |
XM_017024769.1 | XP_016880258.1 | 20 - 20 | 2928 | 99 | |
XM_017024770.1 | XP_016880259.1 | 21 - 21 | 2882 | 99 | |
XM_017024771.1 | XP_016880260.1 | 20 - 20 | 2864 | 99 | |
XM_017024772.1 | XP_016880261.1 | 21 - 21 | 2901 | 99 | |
XM_017024773.1 | XP_016880262.1 | 21 - 21 | 3482 | 99 | |
XM_017024774.1 | XP_016880263.1 | 21 - 21 | 3064 | 99 | |
GenBank mRNA | AK098213.1 | - | 2 - 2 | 828 | 99 |
AK125927.1 | - | 1 - 1 | 3601 | 99 | |
AL096733.1 | - | 1 - 1 | 51 | 99 | |
AL137683.1 | - | 4 - 4 | 1084 | 99 | |
BC032398.1 | - | 22 - 22 | 3020 | 99 | |
BX648978.1 | - | 21 - 21 | 3053 | 99 | |
CR627443.1 | - | 15 - 15 | 1996 | 99 |