The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130782.2 | NP_570138.1 | 4 - 5 | 622 | 116 |
XM_011509891.2 | XP_011508193.1 | 2 - 3 | 189 | 116 | |
GenBank mRNA | AF076642.1 | - | 4 - 5 | 596 | 116 |
AF268036.1 | - | 4 - 5 | 608 | 116 | |
AK057114.1 | - | 4 - 5 | 615 | 116 | |
AK292015.1 | - | 4 - 5 | 614 | 116 | |
AK315377.1 | - | 4 - 5 | 615 | 116 | |
BC020632.1 | - | 4 - 5 | 614 | 116 |