The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001309516.1 | NP_001296445.1 | 13 - 13 | 1702 | 112 |
NM_005048.3 | NP_005039.1 | 13 - 13 | 1598 | 112 | |
GenBank mRNA | AK294181.1 | - | 13 - 13 | 1702 | 112 |
AY497546.1 | - | 13 - 13 | 1384 | 112 | |
BC036811.1 | - | 13 - 13 | 1597 | 112 | |
U25128.1 | - | 13 - 13 | 1526 | 112 |