The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001223.4 | NP_001214.1 | 3 - 4 | 434 | 77 |
NM_001257118.2 | NP_001244047.1 | 4 - 5 | 497 | 77 | |
NM_001257119.2 | NP_001244048.1 | 3 - 4 | 434 | 77 | |
NM_033292.3 | NP_150634.1 | 4 - 5 | 497 | 77 | |
NM_033293.3 | NP_150635.1 | 3 - 4 | 218 | 77 | |
NM_033294.3 | NP_150636.1 | 3 - 4 | 218 | 77 | |
XM_017018393.1 | XP_016873882.1 | 4 - 5 | 845 | 77 | |
XM_017018394.1 | XP_016873883.1 | 4 - 5 | 845 | 77 | |
XM_017018395.1 | XP_016873884.1 | 3 - 4 | 782 | 77 | |
XM_017018396.1 | XP_016873885.1 | 4 - 5 | 754 | 77 | |
GenBank mRNA | AK223503.1 | - | 4 - 5 | 489 | 77 |
AK290114.1 | - | 4 - 5 | 488 | 77 | |
AK290122.1 | - | 3 - 4 | 425 | 77 | |
AK301037.1 | - | 4 - 5 | 397 | 77 | |
AK313516.1 | - | 4 - 5 | 483 | 77 | |
AY660536.1 | - | 4 - 5 | 391 | 77 | |
BC041689.2 | - | 2 - 3 | 383 | 77 | |
BC062327.1 | - | 3 - 4 | 184 | 77 | |
M87507.1 | - | 4 - 5 | 470 | 77 | |
U13697.1 | - | 2 - 3 | 390 | 77 | |
U13698.1 | - | 2 - 3 | 174 | 77 | |
U13699.1 | - | 2 - 3 | 174 | 77 | |
X65019.1 | - | 3 - 4 | 460 | 77 |