The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001944.2 | NP_001935.2 | 13 - 14 | 2145 | 92 |
XM_011525850.1 | XP_011524152.1 | 13 - 14 | 2145 | 92 | |
GenBank mRNA | AK290367.1 | - | 13 - 14 | 2145 | 92 |
M76482.1 | - | 13 - 14 | 2119 | 92 |