Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPN
Entrez Gene ID
3249
Gene Name
hepsin
Gene Aliases
TMPRSS1
Chromosome Location
Chr.19: 35040506 - 35066573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002151.2 NP_002142.1 8 - 9 696 62
NM_182983.2 NP_892028.1 7 - 8 1276 62
XM_005258838.4 XP_005258895.2 7 - 8 774 62
XM_006723181.3 XP_006723244.2 7 - 8 650 62
XM_017026731.1 XP_016882220.1 7 - 8 949 62
XM_017026732.1 XP_016882221.1 6 - 7 511 62
GenBank mRNA AK293622.1 - 6 - 7 530 62
AK315655.1 - 7 - 8 629 62
BC025716.1 - 7 - 8 648 62
EU644753.1 - 6 - 7 451 62
M18930.1 - 8 - 9 696 62
X07002.1 - 2 - 3 112 62
X07732.1 - 7 - 8 1276 62

View Full Product Details