The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317985.1 | NP_001304914.1 | 1 - 2 | 362 | 75 |
NM_001317986.1 | NP_001304915.1 | 1 - 2 | 362 | 75 | |
NM_005702.3 | NP_005693.1 | 1 - 2 | 362 | 75 | |
NR_134328.1 | 1 - 2 | 362 | 75 | ||
GenBank mRNA | AB049388.1 | - | 1 - 2 | 310 | 75 |
AF082657.1 | - | 1 - 2 | 303 | 75 | |
AF082658.1 | - | 1 - 2 | 303 | 75 | |
AK023342.1 | - | 1 - 2 | 303 | 75 | |
AY007435.1 | - | 1 - 2 | 305 | 75 | |
BC019094.2 | - | 1 - 2 | 294 | 75 | |
BI758329.1 | - | 1 - 2 | 273 | 75 | |
BX384612.2 | - | 1 - 2 | 284 | 75 | |
BX537542.1 | - | 1 - 2 | 302 | 75 | |
DA559976.1 | - | 1 - 2 | 363 | 75 |