Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATE1
Entrez Gene ID
11101
Gene Name
arginyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.10: 121740421 - 121928801 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001976.2 NP_001001976.1 1 - 2 216 97
NM_001288734.1 NP_001275663.1 1 - 2 216 97
NM_007041.3 NP_008972.2 1 - 2 216 97
XM_005269458.2 XP_005269515.1 1 - 2 228 97
XM_017015599.1 XP_016871088.1 1 - 2 228 97
XM_017015600.1 XP_016871089.1 1 - 2 54 97
XM_017015602.1 XP_016871091.1 1 - 2 100 97
XM_017015604.1 XP_016871093.1 1 - 2 174 97
GenBank mRNA AF079098.1 - 1 - 2 90 97
AF079099.1 - 1 - 2 90 97
AK296356.1 - 1 - 2 54 97
BC022026.1 - 1 - 2 189 97

View Full Product Details