The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022003.3 | NP_071286.1 | 1 - 2 | 440 | 68 |
GenBank mRNA | AK289629.1 | - | 1 - 2 | 380 | 68 |
AK311736.1 | - | 1 - 2 | 52 | 68 | |
AL035858.1 | - | 1 - 2 | 64 | 68 | |
AY358976.1 | - | 1 - 2 | 50 | 68 | |
BC093040.1 | - | 1 - 2 | 50 | 68 | |
DA209740.1 | - | 1 - 2 | 441 | 68 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204268.1 | NP_001191197.1 | 1 - 2 | 440 | 68 |
NM_001243598.2 | NP_001230527.1 | 1 - 2 | 440 | 68 | |
GenBank mRNA | CN310658.1 | - | 1 - 2 | 75 | 68 |