The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153634.2 | NP_705898.1 | 14 - 15 | 1048 | 106 |
XM_011537951.2 | XP_011536253.1 | 14 - 15 | 4023 | 106 | |
XM_017018851.1 | XP_016874340.1 | 15 - 16 | 2021 | 106 | |
XM_017018852.1 | XP_016874341.1 | 14 - 15 | 994 | 106 | |
GenBank mRNA | AK094998.1 | - | 2 - 3 | 622 | 106 |
AK098593.1 | - | 14 - 15 | 979 | 106 | |
AK307908.1 | - | 14 - 15 | 1021 | 106 | |
BC046366.1 | - | 2 - 3 | 365 | 106 | |
BC110577.1 | - | 14 - 15 | 1039 | 106 | |
BC110578.1 | - | 14 - 15 | 1039 | 106 | |
BX538055.1 | - | 14 - 15 | 1062 | 106 |