Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNHG6
Entrez Gene ID
641638
Gene Name
small nucleolar RNA host gene 6
Gene Aliases
HBII-276HG, NCRNA00058, U87HG
Chromosome Location
Chr.8: 66921930 - 66925542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002599.1 1 - 2 102 63
GenBank mRNA AY264285.1 - 1 - 2 102 63

View Full Product Details