The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014967.4 | NP_055782.3 | 12 - 13 | 3077 | 81 |
XM_005254232.4 | XP_005254289.1 | 12 - 13 | 3085 | 81 | |
XM_005254234.4 | XP_005254291.1 | 12 - 13 | 3131 | 81 | |
XM_005254235.3 | XP_005254292.1 | 12 - 13 | 2967 | 81 | |
XM_011521370.2 | XP_011519672.1 | 12 - 13 | 1770 | 81 | |
XM_011521371.1 | XP_011519673.1 | 13 - 14 | 3196 | 81 | |
XM_017022012.1 | XP_016877501.1 | 11 - 12 | 2847 | 81 | |
XM_017022013.1 | XP_016877502.1 | 11 - 12 | 2860 | 81 | |
GenBank mRNA | AB023235.2 | - | 12 - 13 | 3024 | 81 |
CR936727.1 | - | 13 - 14 | 3191 | 81 | |
CR936734.1 | - | 12 - 13 | 3047 | 81 | |
HM560957.1 | - | 11 - 12 | 2786 | 81 | |
KJ535046.1 | - | 2 - 3 | 751 | 81 |