Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11949377-11949896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLOD1
Entrez Gene ID
5351
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Aliases
EDS6, LH, LH1, LLH, PLOD
UniGene
Hs.75093
Gene Chromosome Location
Chr.1: 11934667-11975542 on Build GRCh38
Transcripts
NM_000302.3 NM_001316320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1333204 c.286A>G p.I96V Chr.1: 11949890-11949890 on Build GRCh38
5018575 c.347G>A p.G116E Chr.1: 11949810-11949810 on Build GRCh38
458663 c.243G>A p.L81L Chr.1: 11949847-11949847 on Build GRCh38
6750259 c.212C>T p.S71L Chr.1: 11949816-11949816 on Build GRCh38
4849176 c.187G>A p.D63N Chr.1: 11949791-11949791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774534534 Chr.1: 11949747-11949747 on Build GRCh38
rs768382193 Chr.1: 11949658-11949658 on Build GRCh38
rs767791340 Chr.1: 11949748-11949748 on Build GRCh38
rs745855996 Chr.1: 11949463-11949463 on Build GRCh38
rs886039207 Chr.1: 11949869-11949869 on Build GRCh38
rs530472841 Chr.1: 11949462-11949462 on Build GRCh38
rs760584840 Chr.1: 11949762-11949762 on Build GRCh38
rs760785117 Chr.1: 11949826-11949826 on Build GRCh38
rs958504190 Chr.1: 11949526-11949526 on Build GRCh38
rs558601008 Chr.1: 11949690-11949690 on Build GRCh38
rs1041674020 Chr.1: 11949635-11949635 on Build GRCh38
rs766073845 Chr.1: 11949767-11949767 on Build GRCh38
rs925589880 Chr.1: 11949646-11949646 on Build GRCh38
rs996356866 Chr.1: 11949418-11949418 on Build GRCh38
rs143604754 Chr.1: 11949797-11949797 on Build GRCh38
rs1024555330 Chr.1: 11949861-11949861 on Build GRCh38
rs371574381 Chr.1: 11949847-11949847 on Build GRCh38
rs944780186 Chr.1: 11949536-11949536 on Build GRCh38
rs1034357016 Chr.1: 11949822-11949822 on Build GRCh38
rs750189989 Chr.1: 11949759-11949759 on Build GRCh38
rs894693033 Chr.1: 11949653-11949653 on Build GRCh38
rs58469200 Chr.1: 11949677-11949677 on Build GRCh38
rs899600120 Chr.1: 11949680-11949680 on Build GRCh38
rs772274349 Chr.1: 11949823-11949823 on Build GRCh38
rs780253192 Chr.1: 11949813-11949813 on Build GRCh38
rs765098815 Chr.1: 11949856-11949856 on Build GRCh38
rs371277921 Chr.1: 11949416-11949416 on Build GRCh38
rs769647081 Chr.1: 11949404-11949404 on Build GRCh38
rs547953454 Chr.1: 11949546-11949546 on Build GRCh38
rs868767291 Chr.1: 11949840-11949840 on Build GRCh38
rs774587722 Chr.1: 11949820-11949820 on Build GRCh38
rs994884283 Chr.1: 11949616-11949616 on Build GRCh38
rs773540306 Chr.1: 11949824-11949824 on Build GRCh38
rs748173461 Chr.1: 11949821-11949821 on Build GRCh38
rs935708407 Chr.1: 11949669-11949669 on Build GRCh38
rs569724013 Chr.1: 11949681-11949681 on Build GRCh38
rs779930427 Chr.1: 11949483-11949483 on Build GRCh38
rs776126385 Chr.1: 11949741-11949741 on Build GRCh38
rs34878020 Chr.1: 11949854-11949854 on Build GRCh38
rs753862253 Chr.1: 11949774-11949774 on Build GRCh38
rs927410990 Chr.1: 11949524-11949524 on Build GRCh38
rs146092290 Chr.1: 11949788-11949788 on Build GRCh38
rs886045200 Chr.1: 11949839-11949839 on Build GRCh38
rs769238622 Chr.1: 11949817-11949817 on Build GRCh38
rs530129382 Chr.1: 11949671-11949671 on Build GRCh38
rs563036918 Chr.1: 11949468-11949468 on Build GRCh38
rs138106022 Chr.1: 11949780-11949780 on Build GRCh38
rs956565246 Chr.1: 11949719-11949719 on Build GRCh38
rs114595968 Chr.1: 11949720-11949720 on Build GRCh38
rs1050177326 Chr.1: 11949495-11949495 on Build GRCh38
rs373927410 Chr.1: 11949783-11949783 on Build GRCh38
rs7541079 Chr.1: 11949871-11949871 on Build GRCh38
rs201579526 Chr.1: 11949841-11949841 on Build GRCh38
rs756553633 Chr.1: 11949804-11949804 on Build GRCh38
rs956421482 Chr.1: 11949447-11949447 on Build GRCh38
rs7551068 Chr.1: 11949805-11949805 on Build GRCh38
rs754877185 Chr.1: 11949777-11949777 on Build GRCh38
rs914893765 Chr.1: 11949722-11949722 on Build GRCh38
rs534551811 Chr.1: 11949731-11949731 on Build GRCh38
rs537417768 Chr.1: 11949689-11949689 on Build GRCh38
rs532620953 Chr.1: 11949511-11949511 on Build GRCh38
rs963451461 Chr.1: 11949489-11949489 on Build GRCh38
rs186778400 Chr.1: 11949585-11949585 on Build GRCh38
rs1005544371 Chr.1: 11949736-11949736 on Build GRCh38
rs749845776 Chr.1: 11949814-11949814 on Build GRCh38
rs375895802 Chr.1: 11949482-11949482 on Build GRCh38
rs766236123 Chr.1: 11949832-11949832 on Build GRCh38
rs563272662 Chr.1: 11949422-11949422 on Build GRCh38
rs536775424 Chr.1: 11949620-11949620 on Build GRCh38
rs61776489 Chr.1: 11949405-11949405 on Build GRCh38
rs897818905 Chr.1: 11949591-11949591 on Build GRCh38
rs34032489 Chr.1: 11949781-11949781 on Build GRCh38
rs77138768 Chr.1: 11949401-11949401 on Build GRCh38
rs945945073 Chr.1: 11949610-11949610 on Build GRCh38
rs923588156 Chr.1: 11949436-11949436 on Build GRCh38
rs992622370 Chr.1: 11949573-11949573 on Build GRCh38

View Full Product Details