The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287499.1 | NP_001274428.1 | 5 - 6 | 603 | 123 |
NM_001287500.1 | NP_001274429.1 | 4 - 5 | 555 | 123 | |
NM_001287501.1 | NP_001274430.1 | 3 - 4 | 509 | 123 | |
NM_001287502.1 | NP_001274431.1 | 3 - 4 | 509 | 123 | |
NM_152558.4 | NP_689771.3 | 5 - 6 | 603 | 123 | |
XM_006715676.2 | XP_006715739.1 | 3 - 4 | 357 | 123 | |
XM_011515243.1 | XP_011513545.1 | 3 - 4 | 484 | 123 | |
XM_017011902.1 | XP_016867391.1 | 5 - 6 | 665 | 123 | |
XM_017011903.1 | XP_016867392.1 | 5 - 6 | 543 | 123 | |
GenBank mRNA | AB028946.2 | - | 3 - 4 | 123 | |
AK092404.1 | - | 5 - 6 | 497 | 123 | |
AK293372.1 | - | 4 - 5 | 378 | 123 | |
AK302053.1 | - | 3 - 4 | 457 | 123 | |
AK309646.1 | - | 5 - 6 | 123 | ||
AL136792.1 | - | 4 - 5 | 284 | 123 | |
BC043150.1 | - | 3 - 4 | 429 | 123 | |
BC061518.1 | - | 4 - 5 | 439 | 123 | |
BC071858.1 | - | 5 - 6 | 577 | 123 | |
KJ534872.1 | - | 5 - 6 | 393 | 123 | |
KJ535081.1 | - | 3 - 4 | 299 | 123 |