Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAB21L3
Entrez Gene ID
126868
Gene Name
mab-21 like 3
Gene Aliases
C1orf161
Chromosome Location
Chr.1: 116072744 - 116135240 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152367.2 NP_689580.2 2 - 3 316 70
XM_011540623.2 XP_011538925.1 2 - 3 448 70
XM_011540625.2 XP_011538927.1 3 - 4 544 70
XM_011540626.2 XP_011538928.1 2 - 3 370 70
XM_017000260.1 XP_016855749.1 3 - 4 1781 70
GenBank mRNA AK096035.1 - 2 - 3 316 70
BC128148.1 - 1 - 2 210 70
BC128149.1 - 1 - 2 210 70

View Full Product Details