Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIK1
Entrez Gene ID
2897
Gene Name
glutamate ionotropic receptor kainate type subunit 1
Gene Aliases
EAA3, EEA3, GLR5, GLUR5, GluK1, gluR-5
Chromosome Location
Chr.21: 29536933 - 29940052 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000830.4 NP_000821.1 2 - 3 844 110
NM_001320616.1 NP_001307545.1 2 - 3 844 110
NM_001320630.1 NP_001307559.1 2 - 3 844 110
NM_175611.2 NP_783300.1 2 - 3 756 110
XM_005260942.1 XP_005260999.1 2 - 3 462 110
XM_005260944.3 XP_005261001.1 2 - 3 462 110
XM_006723991.1 XP_006724054.1 2 - 3 462 110
GenBank mRNA AJ249208.1 - 2 - 3 417 110
AK296404.1 - 2 - 3 789 110
BC028238.1 - 2 - 3 825 110
L19058.1 - 2 - 3 713 110
U16125.1 - 2 - 3 292 110

View Full Product Details