The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077525.2 | NP_001070993.1 | 10 - 11 | 1118 | 68 |
NM_001077526.2 | NP_001070994.1 | 10 - 11 | 1118 | 68 | |
NM_022485.4 | NP_071930.2 | 10 - 11 | 1118 | 68 | |
XM_005265400.1 | XP_005265457.1 | 9 - 10 | 1016 | 68 | |
XM_011534012.1 | XP_011532314.1 | 9 - 10 | 887 | 68 | |
XM_011534013.1 | XP_011532315.1 | 9 - 10 | 843 | 68 | |
XM_017007034.1 | XP_016862523.1 | 11 - 12 | 1288 | 68 | |
XM_017007035.1 | XP_016862524.1 | 10 - 11 | 1213 | 68 | |
XM_017007036.1 | XP_016862525.1 | 11 - 12 | 1287 | 68 | |
XM_017007037.1 | XP_016862526.1 | 10 - 11 | 1213 | 68 | |
XM_017007038.1 | XP_016862527.1 | 9 - 10 | 1016 | 68 | |
XM_017007039.1 | XP_016862528.1 | 11 - 12 | 1287 | 68 | |
XM_017007040.1 | XP_016862529.1 | 10 - 11 | 1213 | 68 | |
XM_017007041.1 | XP_016862530.1 | 8 - 9 | 812 | 68 | |
XM_017007042.1 | XP_016862531.1 | 9 - 10 | 1016 | 68 | |
XM_017007043.1 | XP_016862532.1 | 9 - 10 | 887 | 68 | |
XM_017007044.1 | XP_016862533.1 | 8 - 9 | 934 | 68 | |
XM_017007045.1 | XP_016862534.1 | 8 - 9 | 923 | 68 | |
GenBank mRNA | AF289603.1 | - | 10 - 11 | 1081 | 68 |
AK026058.1 | - | 9 - 10 | 685 | 68 | |
AK074792.1 | - | 10 - 11 | 1089 | 68 | |
AK298846.1 | - | 5 - 6 | 647 | 68 | |
AL713695.1 | - | 10 - 11 | 1039 | 68 | |
AY007098.1 | - | 7 - 8 | 526 | 68 | |
BC001674.1 | - | 10 - 11 | 1023 | 68 | |
BC025952.1 | - | 10 - 11 | 1068 | 68 | |
BC035690.1 | - | 10 - 11 | 1030 | 68 | |
BC050626.1 | - | 10 - 11 | 1041 | 68 | |
DQ630520.1 | - | 10 - 11 | 967 | 68 |