Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTMR14
Entrez Gene ID
64419
Gene Name
myotubularin related protein 14
Gene Aliases
C3orf29
Chromosome Location
Chr.3: 9649433 - 9702394 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077525.2 NP_001070993.1 11 - 12 1205 65
NM_001077526.2 NP_001070994.1 11 - 12 1205 65
NM_022485.4 NP_071930.2 11 - 12 1205 65
XM_005265400.1 XP_005265457.1 10 - 11 1103 65
XM_011534012.1 XP_011532314.1 10 - 11 974 65
XM_011534013.1 XP_011532315.1 10 - 11 930 65
XM_017007034.1 XP_016862523.1 12 - 13 1375 65
XM_017007035.1 XP_016862524.1 11 - 12 1300 65
XM_017007036.1 XP_016862525.1 12 - 13 1374 65
XM_017007037.1 XP_016862526.1 11 - 12 1300 65
XM_017007038.1 XP_016862527.1 10 - 11 1103 65
XM_017007039.1 XP_016862528.1 12 - 13 1374 65
XM_017007040.1 XP_016862529.1 11 - 12 1300 65
XM_017007041.1 XP_016862530.1 9 - 10 899 65
XM_017007042.1 XP_016862531.1 10 - 11 1103 65
XM_017007043.1 XP_016862532.1 10 - 11 974 65
XM_017007044.1 XP_016862533.1 9 - 10 1021 65
XM_017007045.1 XP_016862534.1 9 - 10 1010 65
GenBank mRNA AF289603.1 - 11 - 12 1168 65
AK026058.1 - 10 - 11 772 65
AK074792.1 - 11 - 12 1176 65
AK298846.1 - 6 - 7 734 65
AL713695.1 - 11 - 12 1126 65
AY007098.1 - 8 - 9 613 65
BC001674.1 - 11 - 12 1110 65
BC025952.1 - 11 - 12 1155 65
BC035690.1 - 11 - 12 1117 65
BC050626.1 - 11 - 12 1128 65
DQ630520.1 - 11 - 12 1054 65

View Full Product Details