The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277.2 | NP_001268.2 | 9 - 10 | 1338 | 72 |
NM_212469.1 | NP_997634.1 | 8 - 9 | 1284 | 72 | |
XM_017017147.1 | XP_016872636.1 | 8 - 9 | 1222 | 72 | |
XM_017017148.1 | XP_016872637.1 | 8 - 9 | 2694 | 72 | |
GenBank mRNA | BC036471.2 | - | 8 - 9 | 1162 | 72 |
D10704.1 | - | 9 - 10 | 1148 | 72 |