Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3BP2
Entrez Gene ID
6452
Gene Name
SH3 domain binding protein 2
Gene Aliases
3BP-2, 3BP2, CRBM, CRPM, RES4-23
Chromosome Location
Chr.4: 2793023 - 2841096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122681.1 NP_001116153.1 7 - 8 712 132
NM_001145855.1 NP_001139327.1 7 - 8 855 132
NM_001145856.1 NP_001139328.1 7 - 8 802 132
NM_003023.4 NP_003014.3 7 - 8 853 132
GenBank mRNA AB000462.1 - 7 - 8 853 132
AB000463.1 - 8 - 9 1032 132
AF000936.1 - 6 - 7 592 132
AK124073.1 - 2 - 3 698 132
AK126537.1 - 2 - 3 1069 132
AK225497.1 - 7 - 8 714 132
AK299996.1 - 7 - 8 771 132
AK312286.1 - 7 - 8 650 132
BC022996.1 - 7 - 8 708 132
U56386.1 - 7 - 8 800 132

View Full Product Details