Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40805057-40805318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A15
Entrez Gene ID
10166
Gene Name
solute carrier family 25 member 15
Gene Aliases
D13S327, HHH, ORC1, ORNT1
UniGene
Hs.646645
Gene Chromosome Location
Chr.13: 40789411-40812460 on Build GRCh38
Transcripts
NM_014252.3 NR_038258.1 NR_038259.1

Gene Symbol
TPTE2P5
Entrez Gene ID
100616668
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 5
Gene Aliases
-
UniGene
Hs.660718
Gene Chromosome Location
Chr.13: 40796985-40921750 on Build GRCh38
Transcripts
NM_014252.3 NR_038258.1 NR_038259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7307076 c.380C>T p.T127M Chr.13: 40805183-40805183 on Build GRCh38
7390273 c.398G>A p.R133Q Chr.13: 40805201-40805201 on Build GRCh38
6210636 c.334G>T p.A112S Chr.13: 40805137-40805137 on Build GRCh38
947391 c.439G>A p.A147T Chr.13: 40805242-40805242 on Build GRCh38
1226098 c.440C>T p.A147V Chr.13: 40805243-40805243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151093794 Chr.13: 40805190-40805190 on Build GRCh38
rs951353049 Chr.13: 40805096-40805096 on Build GRCh38
rs9577152 Chr.13: 40805136-40805136 on Build GRCh38
rs758007220 Chr.13: 40805225-40805225 on Build GRCh38
rs199894905 Chr.13: 40805140-40805140 on Build GRCh38
rs775654159 Chr.13: 40805157-40805157 on Build GRCh38
rs769834226 Chr.13: 40805091-40805091 on Build GRCh38
rs781114752 Chr.13: 40805100-40805101 on Build GRCh38
rs768587846 Chr.13: 40805278-40805278 on Build GRCh38
rs543205273 Chr.13: 40805184-40805184 on Build GRCh38
rs747998866 Chr.13: 40805137-40805137 on Build GRCh38
rs780976780 Chr.13: 40805266-40805266 on Build GRCh38
rs757284575 Chr.13: 40805240-40805240 on Build GRCh38
rs186892167 Chr.13: 40805201-40805201 on Build GRCh38
rs886050240 Chr.13: 40805146-40805146 on Build GRCh38
rs201902280 Chr.13: 40805183-40805183 on Build GRCh38
rs374806769 Chr.13: 40805226-40805226 on Build GRCh38
rs774528430 Chr.13: 40805282-40805282 on Build GRCh38
rs868652043 Chr.13: 40805251-40805251 on Build GRCh38
rs779635040 Chr.13: 40805128-40805128 on Build GRCh38
rs752025971 Chr.13: 40805110-40805110 on Build GRCh38
rs754666431 Chr.13: 40805133-40805133 on Build GRCh38
rs371187770 Chr.13: 40805139-40805139 on Build GRCh38
rs780201405 Chr.13: 40805210-40805210 on Build GRCh38
rs752316567 Chr.13: 40805220-40805220 on Build GRCh38
rs553432772 Chr.13: 40805191-40805191 on Build GRCh38
rs775394895 Chr.13: 40805095-40805095 on Build GRCh38
rs936806358 Chr.13: 40805163-40805163 on Build GRCh38
rs758438178 Chr.13: 40805181-40805182 on Build GRCh38
rs777568380 Chr.13: 40805231-40805231 on Build GRCh38
rs191783131 Chr.13: 40805269-40805269 on Build GRCh38
rs750924937 Chr.13: 40805124-40805124 on Build GRCh38
rs749323553 Chr.13: 40805165-40805165 on Build GRCh38
rs377456199 Chr.13: 40805106-40805106 on Build GRCh38
rs756644906 Chr.13: 40805127-40805127 on Build GRCh38
rs573469049 Chr.13: 40805200-40805200 on Build GRCh38
rs35434090 Chr.13: 40805148-40805148 on Build GRCh38
rs753227136 Chr.13: 40805129-40805129 on Build GRCh38
rs745892996 Chr.13: 40805132-40805133 on Build GRCh38
rs770726226 Chr.13: 40805149-40805149 on Build GRCh38
rs764993422 Chr.13: 40805215-40805215 on Build GRCh38
rs754436560 Chr.13: 40805206-40805206 on Build GRCh38
rs763260189 Chr.13: 40805101-40805101 on Build GRCh38
rs762253875 Chr.13: 40805118-40805118 on Build GRCh38
rs377466500 Chr.13: 40805279-40805279 on Build GRCh38

View Full Product Details