Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11947871-11948144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLOD1
Entrez Gene ID
5351
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Aliases
EDS6, LH, LH1, LLH, PLOD
UniGene
Hs.75093
Gene Chromosome Location
Chr.1: 11934667-11975542 on Build GRCh38
Transcripts
NM_000302.3 NM_001316320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3472222 c.306C>T p.I102I Chr.1: 11948064-11948064 on Build GRCh38
1732961 c.168+1G>A p.? Chr.1: 11948068-11948068 on Build GRCh38
5977845 c.109G>A p.E37K Chr.1: 11948008-11948008 on Build GRCh38
4021253 c.127C>T p.R43C Chr.1: 11948026-11948026 on Build GRCh38
4021255 c.127C>T p.R43C Chr.1: 11948026-11948026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769239877 Chr.1: 11947949-11947949 on Build GRCh38
rs556718115 Chr.1: 11948017-11948017 on Build GRCh38
rs781555795 Chr.1: 11948105-11948105 on Build GRCh38
rs779974775 Chr.1: 11948088-11948088 on Build GRCh38
rs148997434 Chr.1: 11948002-11948002 on Build GRCh38
rs775036250 Chr.1: 11947957-11947957 on Build GRCh38
rs142710681 Chr.1: 11948036-11948036 on Build GRCh38
rs765270896 Chr.1: 11947995-11947995 on Build GRCh38
rs758612717 Chr.1: 11947932-11947932 on Build GRCh38
rs141704997 Chr.1: 11948027-11948027 on Build GRCh38
rs202116614 Chr.1: 11948024-11948024 on Build GRCh38
rs371488160 Chr.1: 11947969-11947969 on Build GRCh38
rs538407894 Chr.1: 11948014-11948014 on Build GRCh38
rs764260254 Chr.1: 11948039-11948039 on Build GRCh38
rs758298824 Chr.1: 11948102-11948102 on Build GRCh38
rs926787989 Chr.1: 11948114-11948114 on Build GRCh38
rs369263247 Chr.1: 11948008-11948008 on Build GRCh38
rs763163866 Chr.1: 11948037-11948037 on Build GRCh38
rs746183003 Chr.1: 11948112-11948112 on Build GRCh38
rs778603432 Chr.1: 11948026-11948026 on Build GRCh38
rs766724096 Chr.1: 11947985-11947985 on Build GRCh38
rs756019949 Chr.1: 11948086-11948086 on Build GRCh38
rs199748181 Chr.1: 11948102-11948102 on Build GRCh38
rs371961536 Chr.1: 11948052-11948052 on Build GRCh38
rs180787304 Chr.1: 11948095-11948095 on Build GRCh38
rs767453933 Chr.1: 11948075-11948075 on Build GRCh38
rs892451074 Chr.1: 11947916-11947916 on Build GRCh38
rs752778368 Chr.1: 11947998-11947998 on Build GRCh38
rs765354146 Chr.1: 11948034-11948034 on Build GRCh38
rs746747050 Chr.1: 11947938-11947938 on Build GRCh38
rs113328616 Chr.1: 11947968-11947968 on Build GRCh38
rs752908624 Chr.1: 11947931-11947931 on Build GRCh38
rs762765040 Chr.1: 11947914-11947914 on Build GRCh38
rs777178486 Chr.1: 11947988-11947988 on Build GRCh38
rs771332500 Chr.1: 11948028-11948028 on Build GRCh38
rs779453447 Chr.1: 11948016-11948016 on Build GRCh38
rs750185258 Chr.1: 11948080-11948080 on Build GRCh38
rs776947403 Chr.1: 11948031-11948031 on Build GRCh38
rs757269731 Chr.1: 11947939-11947939 on Build GRCh38
rs768307244 Chr.1: 11947960-11947960 on Build GRCh38
rs759377043 Chr.1: 11947913-11947913 on Build GRCh38
rs11553679 Chr.1: 11948013-11948013 on Build GRCh38
rs753715117 Chr.1: 11948090-11948090 on Build GRCh38
rs202003686 Chr.1: 11948023-11948023 on Build GRCh38
rs748066312 Chr.1: 11948004-11948004 on Build GRCh38
rs376305279 Chr.1: 11947963-11947963 on Build GRCh38
rs374597380 Chr.1: 11947994-11947994 on Build GRCh38
rs781348363 Chr.1: 11947946-11947946 on Build GRCh38
rs748925361 Chr.1: 11947959-11947959 on Build GRCh38
rs945451140 Chr.1: 11948040-11948040 on Build GRCh38
rs751831236 Chr.1: 11948007-11948007 on Build GRCh38
rs777456504 Chr.1: 11947933-11947933 on Build GRCh38
rs771669974 Chr.1: 11948103-11948103 on Build GRCh38
rs890787649 Chr.1: 11947929-11947929 on Build GRCh38
rs776894307 Chr.1: 11947981-11947981 on Build GRCh38
rs138698098 Chr.1: 11948035-11948035 on Build GRCh38
rs374793932 Chr.1: 11947948-11947948 on Build GRCh38
rs762098276 Chr.1: 11948063-11948063 on Build GRCh38

View Full Product Details