The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173086.4 | NP_775109.2 | 9 - 9 | 1849 | 143 |
GenBank mRNA | AK299478.1 | - | 10 - 10 | 1853 | 143 |
BC110639.1 | - | 9 - 9 | 1849 | 143 | |
L42611.1 | - | 9 - 9 | 1841 | 143 |