Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44407310-44407563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNF4A
Entrez Gene ID
3172
Gene Name
hepatocyte nuclear factor 4 alpha
Gene Aliases
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14
UniGene
Hs.116462
Gene Chromosome Location
Chr.20: 44355801-44432845 on Build GRCh38
Transcripts
NM_000457.4 NM_001030003.2 NM_001030004.2 NM_001258355.1 NM_001287182.1 NM_001287183.1 NM_001287184.1 NM_175914.4 NM_178849.2 NM_178850.2 XM_005260407.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3841075 c.378G>A p.K126K Chr.20: 44407468-44407468 on Build GRCh38
6294031 c.253G>T p.D85Y Chr.20: 44407409-44407409 on Build GRCh38
1713513 c.382G>A p.E128K Chr.20: 44407472-44407472 on Build GRCh38
1681697 c.333G>T p.Q111H Chr.20: 44407423-44407423 on Build GRCh38
1713511 c.382G>A p.E128K Chr.20: 44407472-44407472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371557531 Chr.20: 44407389-44407389 on Build GRCh38
rs913090618 Chr.20: 44407402-44407402 on Build GRCh38
rs759922116 Chr.20: 44407373-44407373 on Build GRCh38
rs112386711 Chr.20: 44407494-44407494 on Build GRCh38
rs772786482 Chr.20: 44407397-44407397 on Build GRCh38
rs779512030 Chr.20: 44407525-44407525 on Build GRCh38
rs768402173 Chr.20: 44407525-44407525 on Build GRCh38
rs1019159742 Chr.20: 44407334-44407334 on Build GRCh38
rs751274258 Chr.20: 44407455-44407455 on Build GRCh38
rs563885374 Chr.20: 44407534-44407534 on Build GRCh38
rs780813696 Chr.20: 44407388-44407388 on Build GRCh38
rs761295403 Chr.20: 44407387-44407387 on Build GRCh38
rs375708199 Chr.20: 44407485-44407485 on Build GRCh38
rs368131271 Chr.20: 44407511-44407511 on Build GRCh38
rs6124640 Chr.20: 44407516-44407516 on Build GRCh38
rs376684710 Chr.20: 44407348-44407348 on Build GRCh38
rs937211225 Chr.20: 44407500-44407500 on Build GRCh38
rs193922471 Chr.20: 44407437-44407438 on Build GRCh38
rs896163918 Chr.20: 44407490-44407490 on Build GRCh38
rs766365045 Chr.20: 44407447-44407447 on Build GRCh38
rs3204261 Chr.20: 44407408-44407408 on Build GRCh38
rs1032617027 Chr.20: 44407365-44407365 on Build GRCh38
rs756409703 Chr.20: 44407493-44407493 on Build GRCh38
rs770043312 Chr.20: 44407333-44407333 on Build GRCh38
rs879328773 Chr.20: 44407489-44407489 on Build GRCh38
rs774527972 Chr.20: 44407357-44407357 on Build GRCh38
rs767826185 Chr.20: 44407482-44407482 on Build GRCh38
rs373697693 Chr.20: 44407506-44407506 on Build GRCh38
rs964899181 Chr.20: 44407346-44407346 on Build GRCh38
rs934955113 Chr.20: 44407484-44407484 on Build GRCh38
rs373320902 Chr.20: 44407456-44407456 on Build GRCh38
rs749690709 Chr.20: 44407355-44407355 on Build GRCh38
rs748208417 Chr.20: 44407330-44407330 on Build GRCh38
rs142116849 Chr.20: 44407471-44407471 on Build GRCh38
rs200071662 Chr.20: 44407356-44407356 on Build GRCh38
rs776310756 Chr.20: 44407384-44407384 on Build GRCh38

View Full Product Details