Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NONO
Entrez Gene ID
4841
Gene Name
non-POU domain containing, octamer-binding
Gene Aliases
MRXS34, NMT55, NRB54, P54, P54NRB, PPP1R114
Chromosome Location
Chr.X: 71283192 - 71301168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145408.1 NP_001138880.1 9 - 10 1741 235
NM_001145409.1 NP_001138881.1 7 - 8 1570 235
NM_001145410.1 NP_001138882.1 6 - 7 1407 235
NM_007363.4 NP_031389.3 8 - 9 1627 235
GenBank mRNA AK291140.1 - 8 - 9 1167 235
AK297144.1 - 6 - 7 964 235
BC002364.1 - 8 - 9 1152 235
BC003129.1 - 9 - 10 1278 235
BC012141.1 - 7 - 8 1092 235
BC025280.1 - 8 - 9 1146 235
BC025352.1 - 8 - 9 1149 235
BC028299.1 - 8 - 9 1150 235
BC069616.1 - 7 - 8 1105 235
BC069639.1 - 7 - 8 1105 235
CR456761.1 - 6 - 7 1031 235
KU178234.1 - 6 - 7 1031 235
KU178235.1 - 7 - 8 847 235
L14599.1 - 8 - 9 1167 235
U89867.1 - 8 - 9 1116 235

View Full Product Details