Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCC6
- Entrez Gene ID
-
368
- Gene Name
- ATP binding cassette subfamily C member 6
- Gene Aliases
-
ABC34, ARA, EST349056, GACI2, MLP1, MOAT-E, MOATE, MRP6, PXE, PXE1, URG7
- Chromosome Location
-
Chr.16: 16149565 - 16223616 on Build GRCh38
- Gene Symbol
- ABCC6P2
- Entrez Gene ID
-
730013
- Gene Name
- ATP binding cassette subfamily C member 6 pseudogene 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 14822432 - 14824702 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_023387.1 |
|
2 - 2 |
361 |
102 |
View Full Product Details