Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FUNDC2
- Entrez Gene ID
-
65991
- Gene Name
- FUN14 domain containing 2
- Gene Aliases
-
DC44, HCBP6, HCC3, PD03104
- Chromosome Location
-
Chr.X: 155026789 - 155056916 on Build GRCh38
- Gene Symbol
- F8
- Entrez Gene ID
-
2157
- Gene Name
- coagulation factor VIII
- Gene Aliases
-
AHF, DXS1253E, F8B, F8C, FVIII, HEMA
- Chromosome Location
-
Chr.X: 154835788 - 155022723 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK295164.1 |
- |
1 - 1 |
|
82 |
View Full Product Details