Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL22A1
Entrez Gene ID
169044
Gene Name
collagen type XXII alpha 1
Gene Aliases
-
Chromosome Location
Chr.8: 138588235 - 138914087 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152888.2 NP_690848.1 45 - 46 3794 120
XM_011516883.2 XP_011515185.1 44 - 45 3812 120
XM_011516884.2 XP_011515186.1 43 - 44 3773 120
XM_011516885.2 XP_011515187.1 43 - 44 3758 120
XM_011516886.2 XP_011515188.1 45 - 46 3724 120
XM_011516887.2 XP_011515189.1 41 - 42 2692 120
XM_011516888.2 XP_011515190.1 44 - 45 3815 120
XM_011516889.2 XP_011515191.1 34 - 35 1933 120
XM_017013150.1 XP_016868639.1 45 - 46 3685 120
XM_017013151.1 XP_016868640.1 41 - 42 3650 120
XM_017013152.1 XP_016868641.1 40 - 41 2682 120
GenBank mRNA AF406780.1 - 45 - 46 3781 120
BC042075.1 - 19 - 20 1019 120
BC144535.1 - 33 - 34 2452 120

View Full Product Details