The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152888.2 | NP_690848.1 | 56 - 57 | 4445 | 108 |
XM_011516883.2 | XP_011515185.1 | 55 - 56 | 4463 | 108 | |
XM_011516884.2 | XP_011515186.1 | 54 - 55 | 4424 | 108 | |
XM_011516885.2 | XP_011515187.1 | 54 - 55 | 4409 | 108 | |
XM_011516886.2 | XP_011515188.1 | 56 - 57 | 4375 | 108 | |
XM_011516887.2 | XP_011515189.1 | 52 - 53 | 3343 | 108 | |
XM_011516889.2 | XP_011515191.1 | 45 - 46 | 2584 | 108 | |
XM_017013150.1 | XP_016868639.1 | 56 - 57 | 4336 | 108 | |
XM_017013151.1 | XP_016868640.1 | 52 - 53 | 4301 | 108 | |
XM_017013152.1 | XP_016868641.1 | 51 - 52 | 3333 | 108 | |
GenBank mRNA | AF406780.1 | - | 56 - 57 | 4432 | 108 |
BC042075.1 | - | 30 - 31 | 1670 | 108 | |
BC144535.1 | - | 44 - 45 | 3103 | 108 |