The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197260.1 | NP_001184189.1 | 2 - 3 | 453 | 60 |
NM_001318866.1 | NP_001305795.1 | 2 - 3 | 953 | 60 | |
NM_001318867.1 | NP_001305796.1 | 2 - 3 | 998 | 60 | |
NM_001318868.1 | NP_001305797.1 | 2 - 3 | 955 | 60 | |
NM_001381.4 | NP_001372.1 | 2 - 3 | 955 | 60 | |
XM_017003528.1 | XP_016859017.1 | 2 - 3 | 422 | 60 | |
GenBank mRNA | AF035299.1 | - | 2 - 3 | 446 | 60 |
AF180527.1 | - | 2 - 3 | 360 | 60 | |
AK055944.1 | - | 2 - 3 | 955 | 60 | |
AK127127.1 | - | 2 - 3 | 197 | 60 | |
AK127656.1 | - | 2 - 3 | 197 | 60 | |
AK222883.1 | - | 2 - 3 | 394 | 60 | |
AK296154.1 | - | 2 - 3 | 408 | 60 | |
AK300097.1 | - | 2 - 3 | 360 | 60 | |
AK314678.1 | - | 2 - 3 | 375 | 60 | |
BC111055.1 | - | 2 - 3 | 384 | 60 | |
BC114440.1 | - | 2 - 3 | 381 | 60 | |
DA382785.1 | - | 2 - 3 | 453 | 60 | |
U70987.1 | - | 2 - 3 | 382 | 60 |