The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000601.5 | NP_000592.3 | 1 - 2 | 315 | 132 |
NM_001010931.2 | NP_001010931.1 | 1 - 2 | 315 | 132 | |
NM_001010932.2 | NP_001010932.1 | 1 - 2 | 315 | 132 | |
NM_001010933.2 | NP_001010933.1 | 1 - 2 | 315 | 132 | |
NM_001010934.2 | NP_001010934.1 | 1 - 2 | 315 | 132 | |
XM_006715956.2 | XP_006716019.1 | 2 - 3 | 278 | 132 | |
XM_011516115.2 | XP_011514417.1 | 2 - 3 | 278 | 132 | |
XM_017012097.1 | XP_016867586.1 | 2 - 3 | 278 | 132 | |
XM_017012098.1 | XP_016867587.1 | 2 - 3 | 278 | 132 | |
GenBank mRNA | AB208900.1 | - | 1 - 2 | 222 | 132 |
AK291674.1 | - | 1 - 2 | 250 | 132 | |
AK313694.1 | - | 1 - 2 | 165 | 132 | |
BC022308.1 | - | 1 - 2 | 164 | 132 | |
BC063485.1 | - | 1 - 2 | 173 | 132 | |
BC105797.1 | - | 1 - 2 | 255 | 132 | |
BC130284.1 | - | 1 - 2 | 163 | 132 | |
BC130286.1 | - | 1 - 2 | 163 | 132 | |
BU533235.1 | - | - | 254 | 132 | |
BX495370.1 | - | 1 - 2 | 316 | 132 | |
FJ830861.1 | - | 1 - 2 | 88 | 132 | |
FJ830862.1 | - | 1 - 2 | 88 | 132 | |
L02931.1 | - | 1 - 2 | 148 | 132 | |
M29145.1 | - | 1 - 2 | 189 | 132 | |
M55379.1 | - | 1 - 2 | 88 | 132 | |
M60718.1 | - | 1 - 2 | 160 | 132 | |
M73239.1 | - | 1 - 2 | 153 | 132 | |
M73240.1 | - | 1 - 2 | 153 | 132 | |
M77227.1 | - | 1 - 2 | 170 | 132 | |
U46010.1 | - | 1 - 2 | 142 | 132 | |
X57574.1 | - | 1 - 2 | 165 | 132 |