The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006242.3 | NP_006233.1 | 1 - 1 | 3366 | 96 |
GenBank mRNA | BC047502.1 | - | 1 - 1 | 3366 | 96 |
BX647298.1 | - | 1 - 1 | 1627 | 96 |