Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FOXP1
- Entrez Gene ID
-
27086
- Gene Name
- forkhead box P1
- Gene Aliases
-
12CC4, HSPC215, MFH, QRF1, hFKH1B
- Chromosome Location
-
Chr.3: 70954714 - 71583989 on Build GRCh38
View Full Product Details