Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
Chromosome Location
Chr.2: 96642719 - 96704887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293083.1 NP_001280012.1 14 - 15 1238 93
XM_011512110.2 XP_011510412.1 14 - 15 1255 93
XM_011512111.2 XP_011510413.1 14 - 15 1255 93
XM_011512113.2 XP_011510415.1 14 - 15 1255 93
XM_011512114.2 XP_011510416.1 14 - 15 1255 93
XM_011512116.2 XP_011510418.1 14 - 15 1255 93
XM_011512117.2 XP_011510419.1 14 - 15 1255 93
XM_011512118.2 XP_011510420.1 13 - 14 1135 93
XM_011512119.2 XP_011510421.1 14 - 15 1255 93
XM_011512120.2 XP_011510422.1 13 - 14 1146 93
XM_011512122.2 XP_011510424.1 14 - 15 1256 93
XM_011512125.2 XP_011510427.1 14 - 15 1256 93
XM_011512126.2 XP_011510428.1 14 - 15 1255 93
XM_011512127.2 XP_011510429.1 14 - 15 1256 93
XM_011512128.2 XP_011510430.1 14 - 15 1255 93
XM_017005222.1 XP_016860711.1 14 - 15 1255 93
GenBank mRNA AY461813.1 - 13 - 14 1102 93

View Full Product Details