The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293083.1 | NP_001280012.1 | 3 - 4 | 332 | 66 |
XM_011512110.2 | XP_011510412.1 | 3 - 4 | 349 | 66 | |
XM_011512111.2 | XP_011510413.1 | 3 - 4 | 349 | 66 | |
XM_011512113.2 | XP_011510415.1 | 3 - 4 | 349 | 66 | |
XM_011512114.2 | XP_011510416.1 | 3 - 4 | 349 | 66 | |
XM_011512115.2 | XP_011510417.1 | 3 - 4 | 349 | 66 | |
XM_011512116.2 | XP_011510418.1 | 3 - 4 | 349 | 66 | |
XM_011512117.2 | XP_011510419.1 | 3 - 4 | 349 | 66 | |
XM_011512118.2 | XP_011510420.1 | 3 - 4 | 349 | 66 | |
XM_011512119.2 | XP_011510421.1 | 3 - 4 | 349 | 66 | |
XM_011512122.2 | XP_011510424.1 | 3 - 4 | 350 | 66 | |
XM_011512125.2 | XP_011510427.1 | 3 - 4 | 350 | 66 | |
XM_011512126.2 | XP_011510428.1 | 3 - 4 | 349 | 66 | |
XM_011512127.2 | XP_011510429.1 | 3 - 4 | 350 | 66 | |
XM_011512128.2 | XP_011510430.1 | 3 - 4 | 349 | 66 | |
XM_017005222.1 | XP_016860711.1 | 3 - 4 | 349 | 66 |