Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAHCC1
Entrez Gene ID
57597
Gene Name
BAH domain and coiled-coil containing 1
Gene Aliases
BAHD2
Chromosome Location
Chr.17: 81395431 - 81466332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291324.1 NP_001278253.1 15 - 16 4677 75
XM_011525063.2 XP_011523365.1 16 - 17 5069 75
XM_017024898.1 XP_016880387.1 15 - 16 4728 75
XM_017024899.1 XP_016880388.1 14 - 15 4634 75
GenBank mRNA AB040880.1 - 11 - 12 2428 75

View Full Product Details