Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIZ
Entrez Gene ID
55857
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, HT013, Kizuna, NCRNA00153, PLK1S1, RP69
Chromosome Location
Chr.20: 21125980 - 21246628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163022.1 NP_001156494.1 9 - 10 1752 134
NM_001163023.1 NP_001156495.1 8 - 9 1662 134
NM_001276389.1 NP_001263318.1 8 - 9 1689 134
NM_018474.4 NP_060944.3 10 - 11 1915 134
XM_011529296.2 XP_011527598.1 10 - 11 1918 134
XM_011529297.2 XP_011527599.1 10 - 11 1853 134
XM_011529298.2 XP_011527600.1 10 - 11 1846 134
XM_011529299.2 XP_011527601.1 9 - 10 1747 134
XM_017027951.1 XP_016883440.1 11 - 12 1861 134
XM_017027952.1 XP_016883441.1 9 - 10 1693 134
XM_017027953.1 XP_016883442.1 11 - 12 1933 134
XM_017027954.1 XP_016883443.1 9 - 10 1693 134
GenBank mRNA AF220187.1 - 6 - 7 947 134
AF451990.1 - 10 - 11 1892 134
AK025476.1 - 10 - 11 1846 134
AK293567.1 - 8 - 9 1633 134
AK295355.1 - 9 - 10 1652 134
AL110196.1 - 6 - 7 1140 134
BC039296.1 - 10 - 11 1876 134
BC045826.1 - 9 - 10 1639 134
BC065550.1 - 8 - 9 1496 134
BC090879.2 - 6 - 7 1320 134
BC105093.1 - 10 - 11 1818 134
BC113370.1 - 10 - 11 1818 134

View Full Product Details