The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163022.1 | NP_001156494.1 | 3 - 4 | 375 | 81 |
NM_001276389.1 | NP_001263318.1 | 2 - 3 | 312 | 81 | |
NM_018474.4 | NP_060944.3 | 4 - 5 | 538 | 81 | |
XM_011529296.2 | XP_011527598.1 | 4 - 5 | 541 | 81 | |
XM_011529297.2 | XP_011527599.1 | 4 - 5 | 530 | 81 | |
XM_011529298.2 | XP_011527600.1 | 4 - 5 | 523 | 81 | |
XM_011529299.2 | XP_011527601.1 | 3 - 4 | 370 | 81 | |
XM_017027951.1 | XP_016883440.1 | 5 - 6 | 484 | 81 | |
XM_017027952.1 | XP_016883441.1 | 3 - 4 | 370 | 81 | |
XM_017027953.1 | XP_016883442.1 | 5 - 6 | 556 | 81 | |
XM_017027954.1 | XP_016883443.1 | 3 - 4 | 370 | 81 | |
GenBank mRNA | AF451990.1 | - | 4 - 5 | 81 | |
AK025476.1 | - | 4 - 5 | 469 | 81 | |
AK293567.1 | - | 2 - 3 | 81 | ||
AK295355.1 | - | 3 - 4 | 81 | ||
BC039296.1 | - | 4 - 5 | 81 | ||
BC045826.1 | - | 3 - 4 | 81 | ||
BC065550.1 | - | 2 - 3 | 81 | ||
BC105093.1 | - | 4 - 5 | 81 | ||
BC113370.1 | - | 4 - 5 | 81 | ||
DC386700.1 | - | 3 - 4 | 375 | 81 |