The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134671.2 | NP_001128143.1 | 4 - 5 | 659 | 79 |
NM_024295.5 | NP_077271.1 | 4 - 5 | 659 | 79 | |
XM_006716657.1 | XP_006716720.1 | 3 - 4 | 143 | 79 | |
XM_011517299.1 | XP_011515601.1 | 3 - 4 | 459 | 79 | |
GenBank mRNA | AK023846.1 | - | 3 - 4 | 220 | 79 |
AK124086.1 | - | 3 - 4 | 220 | 79 | |
AY358818.1 | - | 4 - 5 | 537 | 79 | |
BC002457.2 | - | 4 - 5 | 488 | 79 |