Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALNT8
Entrez Gene ID
26290
Gene Name
polypeptide N-acetylgalactosaminyltransferase 8
Gene Aliases
GALNAC-T8
Chromosome Location
Chr.12: 4720586 - 4772726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017417.1 NP_059113.1 10 - 11 1853 82
GenBank mRNA AJ271385.1 - 10 - 11 1853 82
AJ505959.1 - 3 - 4 381 82
BC140888.1 - 10 - 11 1819 82
BC140889.1 - 10 - 11 1819 82

View Full Product Details