Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KPNA4
Entrez Gene ID
3840
Gene Name
karyopherin subunit alpha 4
Gene Aliases
IPOA3, QIP1, SRP3
Chromosome Location
Chr.3: 160494995 - 160565588 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002268.4 NP_002259.1 12 - 13 1339 74
GenBank mRNA AB002533.2 - 12 - 13 1042 74
AK291041.1 - 12 - 13 1291 74
BC028691.2 - 12 - 13 1284 74
BC034493.2 - 12 - 13 1339 74
U93240.1 - 12 - 13 1183 74
Y12393.1 - 3 - 4 313 74

View Full Product Details